Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

m-Toluic acid 98%

Supplier: Ambeed
Warning
Synonyms: 3-Methylbenzoic acid

A107411-500G A107411-1000G A107411-100G
77107-752EA 19.57 USD InStock
77107-752 77107-750 77107-748
m-Toluic acid 98%
m-Toluic acid
Caution: for Research Use Only


Formula: C₈H₈O₂
MW: 136.15 g/mol
MDL Number: MFCD00002523
CAS Number: 99-04-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty