Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Toluic acid 97%

Supplier: Ambeed
Warning
Synonyms: 2-Methylbenzoic acid

A134613-25G A134613-1000G A134613-500G A134613-100G
77109-446EA 24.72 USD InStock
77109-446 77109-444 77109-442 77109-440
o-Toluic acid 97%
o-Toluic acid
Caution: for Research Use Only


Formula: C₈H₈O₂
MW: 136.15 g/mol
MDL Number: MFCD00002477
CAS Number: 118-90-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty