Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Taurine 99+%

Supplier: Ambeed
Warning
Synonyms: 2-Aminoethanesulphonic acid2-Aminoethanesulfonic acid

A184453-500G A184453-25G A184453-100G
77111-524EA 8.24 USD InStock
77111-524 77111-528 77111-526
Taurine 99+%
Taurine
Caution: for Research Use Only


Formula: C₂H₇NO₃S
MW: 125.15 g/mol
MDL Number: MFCD00008197
CAS Number: 107-35-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty