Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Mercaptoethanesulfonic acid sodium salt (mesna) 98%

Supplier: Ambeed
Danger
Synonyms: Sodium 2-mercaptoethanesulfonateSodium 2-mercaptoethanesulphonate

A205146-1MG
77112-296EA 5.15 USD InStock
77112-296
2-Mercaptoethanesulfonic acid sodium salt (mesna) 98%
2-Mercaptoethanesulphonic acid sodium salt (Mesna)
Caution: for Research Use Only


Formula: C₂H₅NaO₃S₂
MW: 164.18 g/mol
MDL Number: MFCD00007535
CAS Number: 19767-45-4
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty