Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-2-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A230218-5G A230218-1G
77112-962EA 9.27 USD InStock
77112-962 77112-964
4-Fluoro-2-nitrobenzonitrile 98%
4-Fluoro-2-nitrobenzonitrile
Caution: for Research Use Only


Formula: C₇H₃FN₂O₂
MDL Number: MFCD00277447
CAS Number: 80517-21-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty