Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Tribromobenzene 97%

Supplier: Ambeed
Warning

A264272-25G A264272-100G A264272-5G A264272-500G A264272-1000G
77113-748EA 5.15 USD InStock
77113-748 77113-746 77113-744 77113-742 77113-740
1,3,5-Tribromobenzene 97%
1,3,5-Tribromobenzene
Caution: for Research Use Only


Formula: C₆H₃Br₃
MW: 314.8 g/mol
MDL Number: MFCD00000080
CAS Number: 626-39-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty