Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triphenylethylene 98%

Supplier: Ambeed
Warning
Synonyms: 1,1,2-Triphenylethylene1,1,2-Triphenylethene

A316020-5G A316020-25G
77114-666EA 88.58 USD InStock
77114-666 77114-668
Triphenylethylene 98%
Triphenylethylene
Caution: for Research Use Only


Formula: (C₆H₅)₂CCHC₆H₅
MW: 256.35 g/mol
MDL Number: MFCD00004765
CAS Number: 58-72-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty