Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

O-Benzylhydroxylamine 98%

Supplier: Ambeed
Danger

A356397-1G A356397-10G A356397-25G
77115-208EA 6.18 USD InStock
77115-208 77115-206 77115-210
O-Benzylhydroxylamine 98%
O-Benzylhydroxylamine
Caution: for Research Use Only


Formula: C₇H₉NO
MDL Number: MFCD00221709
CAS Number: 622-33-3
UN: 2735
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty