Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Difluorophenyl isocyanate 98%

Supplier: Ambeed
Danger
Synonyms: 1,4-Difluoro-2-isocyanatobenzene

A400033-1G
77115-716EA 17.51 USD InStock
77115-716
2,5-Difluorophenyl isocyanate 98%
2,5-Difluorophenyl isocyanate
Caution: for Research Use Only


Formula: C₇H₃F₂NO
MW: 155.1 g/mol
MDL Number: MFCD00013843
CAS Number: 39718-32-6
UN: 3080
ADR: 6.1,3,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty