Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-nitrobenzoate 98%

Supplier: Ambeed
Warning
Synonyms: 4-Nitrobenzoic acid emthyl esterMethyl 4-nitrobenzoate

A427459-25G A427459-500G A427459-100G
77115-998EA 177.16 USD InStock
77115-998 77115-996 77115-994
Methyl-4-nitrobenzoate 98%
Methyl 4-nitrobenzoate
Caution: for Research Use Only


Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD00007350
CAS Number: 619-50-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty