Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitro-α,α,α-trifluorotoluene 97%

Supplier: Ambeed
Danger
Synonyms: 4-Nitro-alpha,alpha,alpha-trifluorotoluene1-Nitro-4-(trifluoromethyl)benzene4-Nitrobenzotrifluoride

A480544-1G
77116-416EA 14.42 USD InStock
77116-416
4-Nitro-α,α,α-trifluorotoluene 97%
4-Nitro-α,α,α-trifluorotoluene
Caution: for Research Use Only


Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
MDL Number: MFCD00007358
CAS Number: 402-54-0
UN: 3431
ADR: 6.1,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty