Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorodiphenylphosphine 98%

Supplier: Ambeed
Danger
Synonyms: DiphenylchlorophosphineDPPC

A489482-25G A489482-10G A489482-5G
77116-464EA 6.18 USD InStock
77116-464 77116-466 77116-468
Chlorodiphenylphosphine 98%
Chlorodiphenylphosphine
Caution: for Research Use Only


Formula: C₁₂H₁₀ClP
MW: 220.64 g/mol
MDL Number: MFCD00000529
CAS Number: 1079-66-9
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty