Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Chlorosuccinimide 97%

Supplier: Ambeed
Danger
Synonyms: 1-Chloropyrrolidine-2,5-dione

A503107-100G A503107-500G A503107-25G A503107-1000G
77116-580EA 27.81 USD InStock
77116-580 77116-582 77116-584 77116-586
N-Chlorosuccinimide 97%
N-Chlorosuccinimide
Caution: for Research Use Only


Formula: C₄H₄ClNO₂
MW: 133.53 g/mol
MDL Number: MFCD00005511
CAS Number: 128-09-6
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty