Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Heptynoic acid 95%

Supplier: Ambeed
Danger

A650746-10G A650746-1G A650746-5G A650746-25G
77117-642EA 40.17 USD InStock
77117-642 77117-640 77117-638 77117-636
6-Heptynoic acid 95%
6-Heptynoic acid
Caution: for Research Use Only


Formula: C₇H₁₀O₂
MDL Number: MFCD00239430
CAS Number: 30964-00-2
UN: 3265
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty