Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning
Synonyms: (3-Nitrophenyl)methanol

A756906-25G A756906-100G A756906-5G A756906-10G
77118-188EA 8.24 USD InStock
77118-188 77118-186 77118-192 77118-190
3-Nitrobenzyl alcohol 98%
3-Nitrobenzyl alcohol
Caution: for Research Use Only


Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD00007273
CAS Number: 619-25-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty