Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,1,3,3,3-Hexafluoro-2-propanol 98%

Supplier: Ambeed
Danger
Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcoholHFIP1,1,1,3,3,3-Hexafluoropropan-2-olHexafluoro-2-propanolHexafluoroisopropanol

A763781-10G A763781-25G
77118-240EA 7.21 USD InStock
77118-240 77118-238
1,1,1,3,3,3-Hexafluoro-2-propanol 98%
1,1,1,3,3,3-Hexafluoro-2-propanol
Caution: for Research Use Only


Formula: C₃H₂F₆O
MW: 168.04 g/mol
MDL Number: MFCD00011651
CAS Number: 920-66-1
UN: 2922
ADR: 8,6.1,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty