Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-DL-alanine 98%

Supplier: Ambeed
Warning
Synonyms: N-Acetyl-DL-2-aminopropionic AcidAc-DL-Ala-OH

A771743-100G A771743-5G A771743-10G
77118-262EA 5.15 USD InStock
77118-262 77118-260 77118-258
N-Acetyl-DL-alanine 98%
N-Acetyl-DL-alanine
Caution: for Research Use Only


Formula: C₅H₉NO₃
MW: 131.13 g/mol
MDL Number: MFCD00037238
CAS Number: 1115-69-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty