Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Histamine (free base) 97%

Supplier: Ambeed
Danger
Synonyms: 2-(1H-Imidazol-4-yl)ethan-1-amine

A804010-100MG A804010-5G A804010-1G
77118-370EA 40.17 USD InStock
77118-370 77118-368
Histamine (free base) 97%
Histamine (free base)
Caution: for Research Use Only


Formula: C₅H₉N₃
MW: 111.15 g/mol
MDL Number: MFCD00005210
CAS Number: 51-45-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty