Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
MitoROS™ 580 Kit for Detecting Reactive Oxygen Species (ROS) in Mitochondria
Supplier: AAT Bioquest
MitoROS™
16052
76482-390EA 240.68 USD InStock
76482-390
MitoROS™ 580 Kit for Detecting Reactive Oxygen Species (ROS) in Mitochondria
Protein Linking and Labeling Systems
Oxidation of MitoROS™ 580 by superoxide generates red fluorescence.
  • Optimized for mitochondria
  • Live cell reagent
MitoROS™ 580 reagent permeates live cells where it selectively targets mitochondria. It is rapidly oxidized by superoxide. It is less likely to be oxidized by other reactive oxygen species (ROS) and reactive nitrogen species (RNS). The oxidized product is highly fluorescent in cells.
Packaging: Plastic vial.
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty