Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-2-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A142939-5G A142939-100G A142939-1G
77137-396EA 33.99 USD InStock
77137-396 77137-392 77137-394
5-Amino-2-nitrobenzoic acid 95%
5-Amino-2-nitrobenzoic acid
Caution: for Research Use Only


Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
MDL Number: MFCD00035748
CAS Number: 13280-60-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty