Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodobenzoic acid 97%

Supplier: Ambeed
Warning
Synonyms: p-Iodobenzoic acid

A146573-100G A146573-10G A146573-5G A146573-25G A146573-500G
77137-574EA 40.17 USD InStock
77137-574 77137-576 77137-578 77137-580 77137-582
4-Iodobenzoic acid 97%
4-Iodobenzoic acid
Caution: for Research Use Only


Formula: C₇H₅IO₂
MW: 248.02 g/mol
MDL Number: MFCD00002533
CAS Number: 619-58-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty