Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloroanisole 98%

Supplier: Ambeed
Warning
Synonyms: 1-Chloro-4-methoxybenzene

A160728-25G A160728-100G A160728-1000G A160728-500G
77138-152EA 10.3 USD InStock
77138-152 77138-150 77138-148 77138-154
4-Chloroanisole 98%
4-Chloroanisole
Caution: for Research Use Only


Formula: C₇H₇ClO
MW: 142.58 g/mol
MDL Number: MFCD00000616
CAS Number: 623-12-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty