Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromobenzoic acid 97%

Supplier: Ambeed
Warning
Synonyms: p-Bromobenzoic Acid

A173445-100G A173445-25G A173445-500G
77138-482EA 6.18 USD InStock
77138-482 77138-480 77138-484
4-Bromobenzoic acid 97%
4-Bromobenzoic acid
Caution: for Research Use Only


Formula: C₇H₅BrO₂
MW: 201.02 g/mol
MDL Number: MFCD00002529
CAS Number: 586-76-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty