Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one 97%

Supplier: Ambeed
Warning

A179257-100MG
77138-646EA 22.66 USD InStock
77138-646
5-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one 97%
5-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one
Caution: for Research Use Only


Formula: C₉H₆ClFO
MW: 184.595 g/mol
MDL Number: MFCD18208183
CAS Number: 1260013-11-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty