Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodobenzene 98%

Supplier: Ambeed
Warning
Synonyms: Phenyliodide

A272695-500G A272695-10G A272695-25G A272695-100G
77141-008EA 99.91 USD InStock
77141-008 77141-002 77141-004 77141-006
Iodobenzene 98%
Iodobenzene
Caution: for Research Use Only


Formula: C₆H₅I
MW: 204.01 g/mol
MDL Number: MFCD00001029
CAS Number: 591-50-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty