Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-2-fluorobenzylamine 97%

Supplier: Ambeed
Danger
Synonyms: (3-Chloro-2-fluorophenyl)methanamine

A423389-1G A423389-5G
77143-192EA 33.99 USD InStock
77143-192 77143-190
3-Chloro-2-fluorobenzylamine 97%
3-Chloro-2-fluorobenzylamine
Caution: for Research Use Only


Formula: C₇H₇ClFN
MW: 159.59 g/mol
MDL Number: MFCD01631391
CAS Number: 72235-55-3
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty