Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichlorobenzamide 98+%

Supplier: Ambeed
Warning

A431846-5G A431846-25G
77143-294EA 43.26 USD InStock
77143-294 77143-296
3,5-Dichlorobenzamide 98+%
3,5-Dichlorobenzamide
Caution: for Research Use Only


Formula: C₇H₅Cl₂NO
MDL Number: MFCD00057644
CAS Number: 5980-23-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty