Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,6-Dichloro-o-toluidine 98%

Supplier: Ambeed
Warning
Synonyms: 2,4-Dichloro-6-methylaniline

A769181-1G A769181-25G A769181-5G
77146-370EA 35.02 USD InStock
77146-370 77146-366 77146-368
4,6-Dichloro-o-toluidine 98%
4,6-Dichloro-o-toluidine
Caution: for Research Use Only


Formula: C₇H₇Cl₂N
MW: 176.05 g/mol
MDL Number: MFCD00044074
CAS Number: 30273-00-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty