Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3,5-dichlorobenzoic acid 97%

Supplier: Ambeed
Warning

A945723-25G A945723-100G A945723-5G
77147-256EA 20.6 USD InStock
77147-256 77147-254 77147-252
4-Amino-3,5-dichlorobenzoic acid 97%
4-Amino-3,5-dichlorobenzoic acid
Caution: for Research Use Only


Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
MDL Number: MFCD00017091
CAS Number: 56961-25-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty