Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Serum Albumin (HSA) Powder-Low Endotoxin, 5 g

Supplier: Gemini Bio-Products
800-126P-005
76994-312EA 307.77 USD InStock
76994-312
Human Serum Albumin (HSA) Powder-Low Endotoxin, 5 g
Sera (Serum)
Human albumin is prepared by cold-alcohol fractionation from pooled human plasma and lyophilized. MW ~66kDa. Endotoxin ≤2 EU/mg.

Delivery information: Ships at ambient. Store refrigerated at 2 to 8 °C

Caution: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty