Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
GoatThroat™ Drum Pump
Supplier: Unimed
GoatThroat™
WGTP520300
77007-802EA 953.02 USD InStock
77007-802
GoatThroat™ Drum Pump
Pumps Drum/Transfer Pumps
Food grade/chemical grade GoatThroat™ Viton® pump with 10.2 cm (4") standoff. GoatThroat™ pumps are ideally suited to transfer disinfectants and sanitizers for COVID-19 from bulk containers into use containers. The GT300 with Viton® seals is the correct pump for hydrogen peroxide, sodium hypochlorite, peroxyacetic acid.
  • Components meet FDA standards
  • Estimated 10 year life expectancy
Food grade/chemical grade GoatThroat™ Viton® pump with 10.2 cm (4") standoff. For use with acids, solvents, herbicides, bio diesel, oils with additives. For viscosities like water, dispensing up to 4 gpm. Flow is good up to 270 centipoise. Includes 1 pump, siphon tube set for up to 106.7 cm (42"), 2" pipe thread fitting fits most barrels, 3 gram packet of molykote 111 grease and single medium drum seal. Note: single drum seal is standard. Other adapters are available for small pails and drums. Made in USA.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty