Would you like to log in again?
Lighthouse Pre-Mix Electric Fog/Mist Unit
Supplier: Unimed
Lighthouse
FPCM516H50
77007-826EA 2190.91 USD InStock
77007-826
Lighthouse Pre-Mix Electric Fog/Mist Unit
Spray Guns
This 120 VAC electric unit lets users apply disinfectants and other chemistry with a handheld wand. With a built-in air compressor, simply plug the unit into a standard wall outlet and get to work.
  • Draws from pre-mixed solution
  • Powered by electricity (120 VAC)
  • Built in air compressor (1.5 hp)
  • 15 micron droplet size at 50 psi (3.4 bar)
  • Compact size for portability
  • Santoprene, Viton, or Kalrez pump seals for chemical compatibility
Unit features a handheld wand with 50' (15.2 m) of twin-line discharge tubing and built-in air compressor (1.5 hp). There is also an option for 100' (30.5 m) of twin-line or 25' (7.6 m) of coiled twin-line discharge tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty