Would you like to log in again?
Lighthouse Foam Unit with Heavy-Duty Trigger Gun, 1.3 Gallon
Supplier: Unimed
Lighthouse
FPUF516PSG
77007-810EA 305.46 USD InStock
77007-810
Lighthouse Foam Unit with Heavy-Duty Trigger Gun, 1.3 Gallon
Spray Guns
This compact unit features a manual pump and is ideal for small jobs and spot cleaning.
  • Maximum portability for small jobs and spot cleaning
  • Pressure relief valve for safety
  • Viton seals for chemical compatibility
  • Natural color tank for easy visibility of product levels
  • Product level fill markings in gallons and liters
  • Extra-wide stability base
This compact unit with manual pump features a heavy-duty trigger gun for additional durability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty