Would you like to log in again?
X-Rite Colorimeter Cell (Lightpaths: 2.5 to 100 mm), Type 96
Supplier: FireFlySci
96G100 96G2.5 96G5 96G20 96G40 96G10
76016-376EA 318.13 USD InStock
76016-376 76016-374 76016-368 76016-366 76016-372 76016-370
X-Rite Colorimeter Cell (Lightpaths: 2.5 to 100 mm), Type 96
Cuvettes Spectrophotometer Cuvettes
Colorimeter cell made from optical glass.
  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in optical glass
  • Lightpaths from 2.5 to 100 mm available
  • Two windows optically polished
Made for X-Rite colorimeters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty