Would you like to log in again?
Circular Dichroism Disc Mount (For Type 42 Cuvette), A26
Supplier: FireFlySci
A26
76016-666EA 216.85 USD InStock
76016-666
Circular Dichroism Disc Mount (For Type 42 Cuvette), A26
Cuvettes Spectrophotometer Cuvettes
Cuvette mount for Type 42 Circular Dichroism Disc Cells.
  • This mount fastens the two sides of the disc together
  • Compatible with Type 42 cuvettes
The A26 Cuvette mount is made from high-quality stainless steel. The mount works by applying pressure to the top release clamp, which opens the sides for insertion of the cuvette.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty