Would you like to log in again?
CardioChek Plus Lipid Panel Sarter Kit
Supplier: PTS Diagnostics
CardioChek Plus
PTS905
77148-110EA 1234.8 USD InStock
77148-110
CardioChek Plus Lipid Panel Sarter Kit
Control reagents
CardioChek Plus lipid panel starter kit provides full lipid panel test results in as little as 90 seconds at the point of care.
Certifications: CLIA-waived, FDA-cleared, Meets NCEP guidelines for accuracy and precision, CRMLN-certified for CHOL and HDL tests.
Ordering information: CardioChek Plus lipid panel starter kit includes: (1) CardioChek Plus analyzer, (4) PTS panels lipid panels (60 lipid tests), (1) PTS panels multi-chemistry controls, (4) PTS collect capillary tubes, 40 µl (64 capillary tubes total).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty