Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A138562-100G A138562-1000G A138562-500G A138562-25G
77315-662EA 5 USD InStock
77315-662 77149-956 77149-954 77149-958
4-Hydroxybenzaldehyde 98%
4-Hydroxybenzaldehyde
Caution: for Research Use Only


Formula: C₇H₆O₂
MW: 122.12 g/mol
MDL Number: MFCD00006939
CAS Number: 123-08-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty