Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylvaleric acid 98%

Supplier: Ambeed
Danger
Synonyms: Isocaproic acid4-Methylpentanoic acid

A335099-25G A335099-5G
77154-792EA 21 USD InStock
77154-792 77154-794
4-Methylvaleric acid 98%
4-Methylvaleric acid
Caution: for Research Use Only


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
MDL Number: MFCD00002803
CAS Number: 646-07-1
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty