Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Dichloro-4-iodobenzene 98%

Supplier: Ambeed
Warning
Synonyms: 3,4-Dichloroiodobenzene

A337746-10G A337746-5G A337746-25G
77154-844EA 8 USD InStock
77154-844 77154-846 77154-848
1,2-Dichloro-4-iodobenzene 98%
1,2-Dichloro-4-iodobenzene
Caution: for Research Use Only


Formula: C₆H₃Cl₂I
MW: 272.9 g/mol
MDL Number: MFCD00019014
CAS Number: 20555-91-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty