Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium formate monohydrate 98%

Supplier: Ambeed
Warning
Synonyms: Formic Acid Lithium SaltLithium formate hydrate

A396746-500G A396746-100G A396746-1000G
77155-574EA 84 USD InStock
77155-574 77155-572 77155-570
Lithium formate monohydrate 98%
Lithium formate monohydrate
Caution: for Research Use Only


Formula: HCO₂Li·H₂O
MW: 51.96 g/mol (anhy)
MDL Number: MFCD00150507
CAS Number: 6108-23-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty