Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-n-Propylheptanoic acid 97%

Supplier: Ambeed
Warning

A405240-250MG A405240-100MG
77155-636EA 41 USD InStock
77155-636 77155-634
2-n-Propylheptanoic acid 97%
2-n-Propylheptanoic acid
Caution: for Research Use Only


Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
MDL Number: MFCD01723758
CAS Number: 31080-39-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty