Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-1,3,5-trichlorobenzene 98%

Supplier: Ambeed
Danger
Synonyms: 1-Bromo-2,4,6-trichlorobenzene

A524166-1G A524166-5G A524166-25G
77156-938EA 21 USD InStock
77156-938 77156-942 77156-940
2-Bromo-1,3,5-trichlorobenzene 98%
2-Bromo-1,3,5-trichlorobenzene
Caution: for Research Use Only


Formula: C₆H₂BrCl₃
MW: 260.34 g/mol
MDL Number: MFCD00060653
CAS Number: 19393-96-5
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty