Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyruvic acid 96%

Supplier: Ambeed
Danger
Synonyms: 2-Oxopropionic acid2-Oxopropanoic acid2-Ketopropionic acidα-Ketopropionic acid

A765171-25G A765171-10G
77158-978EA 5 USD InStock
77158-978 77158-980
Pyruvic acid 96%
Pyruvic acid
Caution: for Research Use Only


Formula: C₃H₄O₃
MW: 88.06 g/mol
MDL Number: MFCD00002585
CAS Number: 127-17-3
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty