Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl 98%

Supplier: Ambeed
Warning
Synonyms: 4-Methacryloyloxy-TEMPO Free RadicalTEMPO MethacrylatePTMA Monomer4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl

A830397-1G
77159-394EA 50 USD InStock
77159-394
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl 98%
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl
Caution: for Research Use Only


Formula: C₁₃H₂₂NO₃
MW: 240.32 g/mol
MDL Number: MFCD17677357
CAS Number: 15051-46-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty