Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orcein (synthetic)

Supplier: Ambeed
Warning
Synonyms: Natural Red 28Orcein (from synthetic)

A874531-100MG A874531-1MG
77159-612EA 12 USD InStock
77159-612 77159-614
Orcein (synthetic)
Orcein
Caution: for Research Use Only


MDL Number: MFCD00062310
CAS Number: 1400-62-0
Order Now
Specification Test Results
Loss on drying max. 25.0 %
Absorbance (E1%1cm) min. 300 (MeOH, 579.0 to 583.0 nm)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty