Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium carbonate 98%

Supplier: Ambeed
Warning
Synonyms: Magnesium Carbonate

A885161-500G A885161-100G
77159-696EA 22 USD InStock
77159-696 77159-694
Magnesium carbonate 98%
Magnesium carbonate
Caution: for Research Use Only


Formula: CMgO₃
MW: 84.31 g/mol
MDL Number: MFCD00064632
CAS Number: 546-93-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty