Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-2,6-difluoroaniline 98%

Supplier: Ambeed
Warning

A896054-250MG A896054-1G
77159-744EA 56 USD InStock
77159-744 77159-746
3-Bromo-2,6-difluoroaniline 98%
3-Bromo-2,6-difluoroaniline
Caution: for Research Use Only


Formula: C₆H₄BrF₂N
MW: 208.006 g/mol
MDL Number: MFCD22627833
CAS Number: 1262198-07-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty