Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Proline 98%

Supplier: Ambeed
Warning
Synonyms: (±)-ProlineH-DL-Pro-OH

A116633-10G A116633-500G A116633-5G A116633-25G A116633-100G
77161-048EA 5 USD InStock
77161-048 77161-046 77161-040 77161-044 77161-042
DL-Proline 98%
DL-Proline
Caution: for Research Use Only


Formula: C₅H₉NO₂
MW: 115.13 g/mol
MDL Number: MFCD00005250
CAS Number: 609-36-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty