Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromoacetaldehyde diethyl acetal 98%

Supplier: Ambeed
Danger
Synonyms: 2-Bromo-1,1-diethoxyethane

A119817-25G A119817-5G
77161-224EA 5 USD InStock
77161-224 77161-222
Bromoacetaldehyde diethyl acetal 98%
Bromoacetaldehyde diethyl acetal
Caution: for Research Use Only


Formula: BrH₂CCH(OCH₂CH₃)₂
MW: 197.07 g/mol
MDL Number: MFCD00000214
CAS Number: 2032-35-1
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty