Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrophthalic anhydride 98%

Supplier: Ambeed
Warning
Synonyms: 5-Nitroisobenzofuran-1,3-dione

A128474-500G A128474-100G A128474-5G A128474-25G
77189-986EA 281 USD InStock
77189-986 77189-988 77189-982 77189-984
4-Nitrophthalic anhydride 98%
4-Nitrophthalic anhydride
Caution: for Research Use Only


Formula: C₈H₃NO₅
MW: 193.12 g/mol
MDL Number: MFCD00005922
CAS Number: 5466-84-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty